missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ETHE1 Monoclonal Antibody (K52048_8E6), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556052

 View more versions of this product

Product Code. 30206992

  • £570.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: RQLSQRGGSG APILLRQMFE PVSCTFTYLL GDRESREAVL IDPVLETAPR DAQLIKELGL RLLYAVNTHC HADHITGSGL LRSLLPGCQS VISRLSGAQA DLHIEDGDSI RFGRFALETR ASPGHTPGCV TFVLNDHSMA FTGDALLIRG CGRTDFQQGC AKTLYHSVHE KIFTLPGDCL IYPAHDYHGF TVSTVEEERT LNPRLTLSCE EFVKIMGNLN

ETHE1 is a sulfur dioxygenase that localizes within the mitochondrial matrix. The enzyme functions in sulfide catabolism. Mutations in its gene result in ethylmalonic encephalopathy.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ETHE1
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
O95571, Q9DCM0
Ethe1
Recombinant human ETHE1 (immunogen range is 11-230).
200 μL
Primary
Human, Mouse
Antibody
IgG1
Western Blot
K52048_8E6
Unconjugated
Ethe1
0610025L15Rik; ETHE1; ETHE1, persulfide dioxygenase; ethylmalonic encephalopathy 1; Ethylmalonic encephalopathy protein 1; Ethylmalonic encephalopathy protein 1 homolog; hepatoma subtracted clone one protein; HSCO; Persulfide dioxygenase ETHE1, mitochondrial; protein ETHE1, mitochondrial; sulfur dioxygenase ETHE1; YF13H12
Mouse
Protein A
RUO
23474, 66071
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.