missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ETHE1 Monoclonal specifically detects ETHE1 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | ETHE1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 81 |
| Conjugate | DyLight 550 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | D83198, EC 3.1.2.6, ethylmalonic encephalopathy 1, Ethylmalonic encephalopathy protein 1, Hepatoma subtracted clone one protein, HSCOEC 3.-, protein ETHE1, mitochondrial, YF13H12 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human ETHE1 (O95571; Leu13-Ala254). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?