Learn More
ETHE1 Antibody (001), Janelia Fluor™ 646, Novus Biologicals™
Product Code. 18408328 Shop All Bio Techne Products
Product Code. 18408328 Supplier Novus Biologicals Supplier No. NBP290300JF646
Rabbit Monoclonal Antibody
ETHE1 Monoclonal antibody specifically detects ETHE1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation