missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ethanolamine kinase Antibody (3F11), Novus Biologicals™

Product Code. 18345799 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345799 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345799 Supplier Novus Biologicals™ Supplier No. H00055500M09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ethanolamine kinase Monoclonal antibody specifically detects ethanolamine kinase in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ethanolamine kinase
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 3F11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_061108
Gene Alias EC 2.7.1.82, EKI, EKI 1, EKI1putative protein product of Nbla10396, ethanolamine kinase 1, Nbla10396
Host Species Mouse
Immunogen ETNK1 (NP_061108, 133 a.a. ∽ 228 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. WDPQEVTLQLFTDGITNKLIGCYVGNTMEDVVLVRIYGNKTELLVDRDEEVKSFRVLQAHGCAPQLYCTFNNGLCYEFIQGEALDPKHVCNPAIFR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55500
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.