missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine-d7, TRC

Product Code. 30386304
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386304 50 mg -
30376156 10 mg 10mg
30386743 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30386304 Supplier Toronto Research Chemicals Supplier No. TRCE67650650MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219802-89-7
Molecular Formula C2 D7 N O
Molecular Weight (g/mol) 68.126
Synonym Ethan-1,1,2,2-d4-ol-d, 2-(amino-d2)- (ACI), 2-(Amino-d2)ethan-1,1,2,2-d4-ol-d (ACI), Ethanolamine-d7, 2-Aminoethanol-D7
IUPAC Name N,N,1,1,2,2-hexadeuterio-2-deuteriooxyethanamine
SMILES [2H]OC([2H])([2H])C([2H])([2H])N([2H])[2H]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C2H7NO/c3-1-2-4/h4H,1-3H2/i1D2,2D2,4D/hD2
Formula Weight 68.097
Chemical Name or Material Ethanolamine-d7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.