missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine, 99%, H2O 0.5% max, Thermo Scientific Chemicals

Product Code. 11358097
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358097 1 kg 1kg
11368097 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358097 Supplier Thermo Scientific Alfa Aesar Supplier No. 022793.A1

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ethanolamine is used as a surfactant, fluorimetric reagent, and to remove CO2 and H2S from natural gas and other gases, functional fluids, intermediates. It is also used as feedstock in the production of detergents, emulsifiers, polishes, pharmaceuticals, corrosion inhibitors, chemical intermediates, ion exchange agents, plating agents and surface treating agents, electrical and electronic products, paper products, personal care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine
PubChem CID 700
ChEBI CHEBI:16000
IUPAC Name 2-aminoethanol
SMILES NCCO

Specifications

Melting Point 10°C
Density 1.012g/mL
Boiling Point 169°C to 170°C
Flash Point 93° (199°F)
Odor Ammonia-like
Refractive Index 1.4540
Quantity 1 kg
UN Number UN2491
Beilstein 505944
Sensitivity Air Sensitive & Hygroscopic
Merck Index 14,3727
Formula Weight 61.08
Percent Purity 99%
Concentration or Composition (by Analyte or Components) H2O 0.5% max.
Physical Form Liquid
Chemical Name or Material Ethanolamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.