missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine, 98+%

Product Code. 11499203
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499203 500 g 500g
11409213 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499203 Supplier Thermo Scientific Alfa Aesar Supplier No. A11697.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141-43-5 | C2H7NO | 61.08 g/mol

Ethanolamine is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethanolamine is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.

Solubility
Miscible with water.

Notes
Store it away from heat, spark, open flames, hot surfaces and smoke. The storage area should be safe and enclosed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine
PubChem CID 700
ChEBI CHEBI:16000
SMILES NCCO

Specifications

Melting Point 10°C
Density 1.012
Boiling Point 169°C to 170°C
Flash Point 93°C (199°F)
Odor Ammonia-like
Linear Formula HOCH2CH2NH2
Refractive Index 1.454
Quantity 500 g
UN Number UN2491
Beilstein 505944
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,3727
Solubility Information Miscible with water.
IUPAC Name 2-aminoethanol
Formula Weight 61.08
Percent Purity ≥98%
Chemical Name or Material Ethanolamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanolamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Ethanolamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.