missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine-13C2 Hydrochloride, TRC

Product Code. 30380899
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380899 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380899 Supplier Toronto Research Chemicals Supplier No. TRCE67650410MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethanolamine-13C2 Hydrochloride
CAS 1173019-25-4
Molecular Formula 13C2H7NO • (HCl)
Quantity 10 mg
Synonym 2-Aminoethanol-13C2 Hydrochloride, 1-Amino-2-hydroxyethane-13C2 Hydrochloride, 2-Amino-1-ethanol-13C2 Hydrochloride, 2-Aminoethanol-13C2 Hydrochloride, 2-Aminoethyl Alcohol-13C2 Hydrochloride
InChI Formula InChI=1S/C2H7NO.ClH/c3-1-2-4;/h4H,1-3H2;1H/i1+1,2+1;
SMILES N[13C][13C]O.Cl
Molecular Weight (g/mol) 63.073646
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.