missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine @ 1000 μg/mL in H2O, VHG™

Product Code. 17979984
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17979984 100 mL 100mL
17948584 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17979984 Supplier VHG Labs Supplier No. VHGIETA1K100
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Ethanolamine @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethanolamine
CAS 141-43-5
Methods Ion Chromatography
Analyte or Component Names Ethanolamine
Shipping Condition Room Temperature
Molecular Formula C2 H7 N O
Synonym C2H7NO, Trolamine Imp. A (EP), 2-Aminoethanol, 1-Amino-2-hydroxyethane, 2-Amino-1-ethanol, 2-Aminoethyl alcohol, 2-Ethanolamine, 2-Hydroxyethanamine, 2-Hydroxyethylamine, Aminoethanol, Colamine, ETA, Envision Conditioner PDD 9020, Ethanolamine, Ethylolamine, Glycinol, Kolamin, MEA, MEA (alcohol), MEA 90, MEA-LCI, Mealan, Monoethanolamine, Olamine, R 100, R 100 (amine), Seramine, beta-Aminoethanol, beta-Aminoethyl alcohol, beta-Ethanolamine, beta-Hydroxyethylamine, Ifosfamide Impurity D, Trolamine Impurity A, 2-Aminoethanol, 1,10-Decanedicarboxylic Acid, 1,10-Dicarboxydecane, 1,12-Dodecanedioic Acid, Corfree M 2, Decamethylenedicarboxylic Acid, NSC 400242, SL-AH, n-Dodecane-α,ω-dioic Acid, n-Dodecanedioic Acid
InChI Formula InChI=1S/C2H7NO/c3-1-2-4/h4H,1-3H2
SMILES NCCO
IUPAC Name 2-aminoethanol
Solvent or Matrix Water
Molecular Weight (g/mol) 61.08
Quantity 100 mL
Formula Weight 61.0528 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.