missing translation for 'onlineSavingsMsg'
Learn More

Ethanol (Untaxed), Riedel-de Haën™

Product Code. 15615715
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Grade:
ACS
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Grade unitSize
15615715 1 L Glass Bottle ACS 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 15615715 Supplier Honeywell-Riedel-de Haen Supplier No. 322211LGL
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Puriss. p.a., absolute, ≥99.8% (GC), for license holders only

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -115°C
CAS 64-17-5
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Molecular Formula C2H6O
Synonym Ethyl alcohol
Boiling Point 78°C to 79°C (1.013 hPa)
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Percent Purity 0.998
Grade ACS
Chemical Name or Material Ethanol (Untaxed)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.