missing translation for 'onlineSavingsMsg'
Learn More

Ethanol (Untaxed), Puriss., absolute, sales not in Germany, ≥99.8% (GC), for license holders only, Honeywell Riedel-de Haën™
BIOPHARMA

Product Code. 15672470
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Poly Bottle
Stainless Steel Can
Unit Size:
1 L
2.5 L
30kg
5 L
Pack of 4
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15672470 2.5 L Poly Bottle 2.5 L
15662470 1 L Poly Bottle 1 L
15612480 5 L Poly Bottle 5 L
15622480 6 x 1 L Poly Bottle Pack of 6
15692470 4 x 2.5 L Poly Bottle Pack of 4
15602480 4 x 5 L Poly Bottle Pack of 4
15682470 30 kg Stainless Steel Can 30kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 15672470 Supplier Honeywell-Riedel-de Haen Supplier No. 241032.5L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C (literature)
CAS Max % 100.0000%
Density 0.789 g/mL at 25 °C(lit.)
Quantity 2.5 L
Flash Point 12°C
Packaging Poly Bottle
Linear Formula CH3CH2OH
UN Number UN1170
Beilstein 1718733
Boiling Point 78°C to 79°C
Formula Weight 46
Percent Purity >99.8%
Grade puriss.
Chemical Name or Material Ethanol (Untaxed), Puriss., absolute, sales not in Germany, ≥99.8% (GC), for license holders only
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.