missing translation for 'onlineSavingsMsg'
Learn More

Solstice Ethanol (Untaxed) CHROMASOLV™

Product Code. 16695723
Change view
Click to view available options
Quantity:
7 L
Unit Size:
7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16695723 7 L 7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 16695723 Supplier Solstice Supplier No. 028557L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Ethanol (Untaxed) CHROMASOLV™ for HPLC, absolute, without additive, A15, ≥99.8%, for license holders only.

  • CHROMASOLV™ for HPLC
  • Absolute, without additive
  • A15
  • ≥99.8%
  • For license holders only

Specifications

pH No data available
Melting Point -115°C
CAS 64-17-5
Color Colorless
Density 0.790 to 0.791 g/cm3 (20°C)
Quantity 7 L
Flash Point 12°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV
Molecular Formula C2H6O
Linear Formula CH3CH2OH
Vapor Pressure 59 hPa (20°C)
MDL Number MFCD00003568
UN Number UN1170
Beilstein 1718733
Synonym Ethyl alcohol
Boiling Point 78°C to 79°C (1.013 hPa)
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Percent Purity ≥99.80%
Grade HPLC
Chemical Name or Material Ethanol (Untaxed)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.