missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol (Taxed), SPECTRANAL™, Absolute, ≥99.8% (GC), for domestic sale in Germany, France, Ireland and United Kingdom, Honeywell Riedel-de Haen
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
Specifications
Specifications
| Melting Point | -114°C (literature) |
| CAS Max % | 100.0000% |
| Density | 0.789 g/mL at 25 °C(lit.) |
| Boiling Point | 78°C to 79°C |
| Quantity | 100 mL |
| Flash Point | 12°C |
| Packaging | Glass Bottle |
| Linear Formula | CH3CH2OH |
| UN Number | UN1170 |
| Beilstein | 1718733 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto