missing translation for 'onlineSavingsMsg'
Learn More

Ethanol (Taxed), CHROMASOLV™, for HPLC, absolute, ≥99.8%, for domestic sale in Germany, France, Ireland and United Kingdom, Honeywell Riedel-de Haen

Product Code. 15624530
Change view
Click to view available options
Quantity:
4 x 2.5 L
Packaging:
Glass Bottle
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15624530 4 x 2.5 L Glass Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15624530 Supplier Honeywell-Chromasolv Supplier No. 348524X2.5L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C (literature)
CAS Max % 100.0000%
Density 0.789 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Flash Point 12°C
Packaging Glass Bottle
Linear Formula CH3CH2OH
UN Number UN1170
Beilstein 1718733
Boiling Point 78°C to 79°C
Formula Weight 46
Percent Purity >99.8%
Grade HPLC
Chemical Name or Material Ethanol (Taxed), CHROMASOLV™, for HPLC, absolute, ≥99.8%, for domestic sale in Germany, France, Ireland and United Kingdom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.