missing translation for 'onlineSavingsMsg'
Learn More

Ethanol solution (Untaxed), Tested according to Ph.Helv., 70 % (v/v) in H2O, for license holders only, Solstice
BIOPHARMA

Product Code. 15650630
Change view
Click to view available options
Quantity:
1 L
25 L
Packaging:
Poly Bottle
Stainless Steel Can
Unit Size:
1 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15650630 25 L Stainless Steel Can 25 L
15630630 1 L Poly Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 15650630 Supplier Honeywell-Riedel-de Haen Supplier No. 0287725L
  • ONLINE EXCLUSIVE PRICE
    £590.00
    £493.15 / 25 L
    Save £96.85
    16% Off
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Min % ≥70.0000%, ≥30.0000%
CAS Max % <90.0000%, <50.0000%
Density 0.886 g/cm3
Quantity 25 L
Flash Point 22°C
Packaging Stainless Steel Can
Linear Formula CH3CH2OH
UN Number UN1170
Beilstein 1718733
Boiling Point 77°C
Molecular Weight (g/mol) 46.07
Chemical Name or Material Ethanol solution (Untaxed), Tested according to Ph.Helv., 70 % (v/v) in H2O, for license holders only
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.