missing translation for 'onlineSavingsMsg'
Learn More

Ethanol solution (Untaxed), Tested according to Ph.Helv., 70 % (v/v) in H2O, for license holders only, Solstice
BIOPHARMA

Product Code. 15630630
Change view
Click to view available options
Quantity:
1 L
25 L
Packaging:
Poly Bottle
Stainless Steel Can
Unit Size:
1 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15630630 1 L Poly Bottle 1 L
15650630 25 L Stainless Steel Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 15630630 Supplier Honeywell-Riedel-de Haen Supplier No. 028771L
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Min % ≥70.0000%, ≥30.0000%
CAS Max % <90.0000%, <50.0000%
Density 0.886 g/cm3
Quantity 1 L
Flash Point 22°C
Packaging Poly Bottle
Linear Formula CH3CH2OH
UN Number UN1170
Beilstein 1718733
Boiling Point 77°C
Molecular Weight (g/mol) 46.069
Chemical Name or Material Ethanol solution (Untaxed), Tested according to Ph.Helv., 70 % (v/v) in H2O, for license holders only
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.