missing translation for 'onlineSavingsMsg'
Learn More

Ethanol Solution 96%, Molecular Biology Grade, Regulated, Fisher BioReagents™

Product Code. p-5155068
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 15552393

Brand: Fisher BioReagents BP8202500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Ideal for concentrating nucleic acid samples by precipitation

  • DNase, and Protease tested to verify absence of hydrolytic enzymes
  • Combine this blend with commercial “mini-prep“ kits or traditional extraction
  • ACS grade Absolute Ethanol used in preparation of solution
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5, 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Colorless
Density 0.80 g/cm3
Boiling Point 78°C
Quantity 500 mL
Packaging HDPE Bottle
Physical Form Liquid
Residue after Evaporation ≤0.001%
Water 2% to 6% (GC to TCD)
Grade Molecular Biology
Chemical Name or Material 96% Ethanol Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.