missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, puriss., 96.0 to 97.2%, Honeywell™
Click to view available options
Quantity:
4 x 2.5 L
4 x 5 L
Unit Size:
Pack of 4
Specifications
Specifications
| Density | 0.805 to 0.812g/mL at 20°C, 0.789g/mL at 25°C |
| Quantity | 4 x 5 L |
| Explosion Limit | 19% (60°F) |
| Flash Point | 14°C (57.2°F) |
| Ignition Point | 683°F |
| Linear Formula | CH3CH2OH |
| Vapor Pressure | 44.6mmHg (20°C) |
| UN Number | UN1170 |
| Beilstein | 1718733 |
| Formula Weight | 46.07g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction