missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, Honeywell™

Product Code. 16685863 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
2.5L GLASS BOTTLE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16685863 2.5L GLASS BOTTLE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 16685863 Supplier Honeywell Research Chemicals Supplier No. 40347H2.5L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Absolute, semiconductor grade PURANAL™ (Honeywell 17833), ≥99.8% (vol.)

  • comprehensive portfolio for the electronics and semiconductor industry
  • for scientists engaged in education, research, development, prototyping and small scale fabrication of high tech micro- and nanochips
  • Superior purity, stability and exceptional lot-to-lot consistency
  • includes high purity products required for high performance applications such as etchants, solvents or hexamethyldisilazane (HMDS)
TRUSTED_SUSTAINABILITY

Specifications

CAS 64-17-5
Quantity 2.5 L
Flash Point 12 °C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3612 - 1.3618
Molecular Formula C2H6O
Water Max. 0.2 %
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
PubChem CID 702
ChEBI CHEBI:16236
Percent Purity min. 99.8 Vol.%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.