missing translation for 'onlineSavingsMsg'
Learn More

Ethanol-d, TRC

Product Code. 30380726
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380726 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380726 Supplier Toronto Research Chemicals Supplier No. TRCE8901232.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethanol-d
CAS 925-93-9
Molecular Formula C2H5DO
Quantity 2.5 g
Synonym Ethyl Alcohol-d, Alcohol-d, Alcohol-d Anhydrous, Algrain-d, Anhydrol-d, Bioethanol-d, Black Warrant-d, CDA 19-d, Denatured Alcohol-d, Denatured Ethanol-d, Desinfektol EL-d, Esumiru WK 88-d, Ethicap-d, Ethyl Hydrate-d, Ethyl Hydroxide-d, Germ-X-d, Hinetoless-d, IMS 99-d, Infinity Pure-d, Jaysol-d, Jaysol S-d, Lux-d, Methylcarbinol-d, Molasses Alcohol-d, NSC 85228-d, Neocol CQ-d, Potato Alcohol-d, SDA 3A-d, SY Fresh M-d, Sekundasprit-d, Sterillium Rub-d, Synasol-d, Tecsol-d, Vinic Alcohol-d
InChI Formula InChI=1S/C2H6O/c1-2-3/h3H,2H2,1H3/i3D
SMILES [2H]OCC
IUPAC Name deuteriooxyethane
Molecular Weight (g/mol) 47.07
Formula Weight 47.0481
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.