missing translation for 'onlineSavingsMsg'
Learn More

Ethanol-d{6}, anhydrous, 99+% (Isotopic)

Product Code. 11337218
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337218 1 g 1g
11347218 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11337218 Supplier Thermo Scientific Alfa Aesar Supplier No. 016799.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

NMR Solvent | CAS: 1516-08-1 | C2H6O | 52.106 g/mol

Ethanol-d_6, is employed as an NMR solvent. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1516-08-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 52.106
MDL Number MFCD00051020
InChI Key LFQSCWFLJHTTHZ-LIDOUZCJSA-N
Synonym ethanol-d6, 2h6 ethanol, hexadeuteroethanol, 2 h? ethan 2 h ol, ethyl-d5 alcohol-d, ethyl alcohol-d6, ethanol d, ethanol-d6 9ci
PubChem CID 102138
IUPAC Name 1,1,1,2,2-pentadeuterio-2-deuteriooxyethane
SMILES CCO

Specifications

Physical Form Liquid
Color Colorless
Density 0.91 g/mL
Odor Alcohol-like
Sensitivity Hygroscopic
Quantity 1 g
Boiling Point 79°C
Linear Formula CD3CD2OD
UN Number UN1170
Solubility Information Soluble in water.
Formula Weight 52.11
Percent Purity ≥99%
Chemical Name or Material Ethanol-d6, Isotopic, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.