Learn More
Ethanol, anhydrous, denatured, 85.8%, 13.3% methanol, 0.9% MIBK, Thermo Scientific Chemicals
£20.10
Chemical Identifiers
CAS | 64-17-5 |
---|---|
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.069 |
MDL Number | MFCD00003568 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Description
Denatured ethanol is acting as a solvent to remove the paint and ink stain on curtains, dress and upholstery. It is utilized as fuel in camping and primus stoves. It is likewise employed to clean the windows, manufacturing nail-polish and to test pectin levels in jam and jelly making.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- As a gasoline additive/substitute; octane booster
- Very mobile liquid solvent
- Used in lotions, tonics and perfumes
Chemical Identifiers
64-17-5 | |
46.069 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |
C2H6O | |
MFCD00003568 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
Specifications
-100°C | |
Colorless | |
80°C | |
Like alcohol | |
1.3610 | |
C2H6O | |
MFCD00003568 | |
1718733 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46.08 | |
13.3% methanol, 0.9% MIBK |
64-17-5 | |
0.811g/mL | |
11° (52°F) | |
Liquid | |
85.8% | |
CH3CH2OH | |
UN1993 | |
14,3760 | |
Miscible in water and most organic solvents | |
CCO | |
46.069 | |
CHEBI:16236 | |
85.8% | |
Ethanol, Anhydrous, denatured |
Safety and Handling
GHS H Statement
H225-H370
Highly flammable liquid and vapor.
Causes damage to organs.
GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'rtecsNumber' : KQ6300000
missing translation for 'tsca' : TSCA
For Manufacturing and Laboratory Use. Not for beverage use