missing translation for 'onlineSavingsMsg'
Learn More

Ethanol absolute, denaturated with 4.8% methanol, A15 METHYL, ≥99.8% (based on denaturant-free substance), Honeywell

Product Code. 15610630 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Plastic Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15610630 1 L Plastic Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 15610630 Supplier Honeywell Research Chemicals Supplier No. 028751L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Specifications

CAS 64-17-5, 67-56-1
CAS Min % ≥50.0000%, ≥3.0000%
CAS Max % ≤100.0000%, <10.0000%
Density 0.78 g/cm3
Quantity 1 L
Flash Point 14°C
Packaging Plastic Bottle
Molecular Formula C2H6O
Linear Formula CH3CH2OH
MDL Number MFCD00003568
UN Number UN1987
Water max. 0.2%
Beilstein 1718733
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Boiling Point 78°C to 80°C
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.069
PubChem CID 702
ChEBI CHEBI:16236
Formula Weight 46
Percent Purity >99.8%
Chemical Name or Material Ethanol absolute, denaturated with 4.8% methanol, A15 METHYL, ≥99.8% (based on denaturant-free substance)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: methanol; ethanol; ethyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.