missing translation for 'onlineSavingsMsg'
Learn More

Ethanol absolute, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. p-7220852 Shop All J.T.Baker Products
Click to view available options
Quantity:
250 mL
2.5 L
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
2.5 L
250mL
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 15568454

Brand: JT Baker 8025.0250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Exceeds ACS Specifications

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Ethyl alcohol
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -117°C
Color Max. 10 (APHA)
Density 0.7895 g/cm³ (20°C)
Boiling Point 78.3°C (1013 hPa)
Quantity 250 mL
Flash Point 12°C
Packaging Glass Bottle
Assay Percent Range min. 99.9% (v/v)
Residue after Evaporation max. 0.001%
Titratable Acid max. 0.0005 meq/g
Titratable Base max. 0.0002 meq/g
UN Number 1170
Water max. 0.2%
Show More Show Less
compliance-icons
Product Identifier
  • Ethanol Absolute
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.