missing translation for 'onlineSavingsMsg'
Learn More

Ethanol absolute, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 15588454 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
250 mL
2.5 L
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
2.5 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15588454 2.5 L Plastic Bottle 2.5 L
15568454 250 mL Glass Bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 15588454 Supplier JT Baker Supplier No. 8025.2500PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Exceeds ACS Specifications

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Ethyl alcohol
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -117°C
Color Max. 10 (APHA)
Density 0.7895 g/cm³ (20°C)
Boiling Point 78.3°C (1013 hPa)
Quantity 2.5 L
Flash Point 12°C
Packaging Plastic Bottle
Assay Percent Range min. 99.9% (v/v)
Residue after Evaporation max. 0.001%
Titratable Acid max. 0.0005 meq/g
Titratable Base max. 0.0002 meq/g
UN Number 1170
Water max. 0.2%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol Absolute
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.