missing translation for 'onlineSavingsMsg'
Learn More

Ethanol Absolute, 99.9%, BAKER ANALYZED™, J.T. Baker™

Catalog Number 13225483
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
HDPE Bottle
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
64-17-5
C2H6O
46.07
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
64-17-5
C2H6O
46.07

Exceeds ACS specifications to provide improved consistency and characterization of the reagent. J.T. Baker™ Ethanol Absolute, 99.9%, BAKER ANALYZED™ meets EPA method 130.2 for testing of water hardness and meets US FDA requirements for use in the pharmaceutical industry.

BAKER ANALYZED™ reagents are a grade of high quality chemicals for laboratory and specialized industrial use. Every BAKER ANALYZED™ Reagent label provides an Actual Lot Analysis as a guarantee of purity and consistency. Analytical methods used for these determinations are based on standard test procedures described in technical publications.

Melting Point −114.5°C
Color Colorless
Percent Purity Min. 99.9% v/v
Density 0.79g/cm3 at 20°C
Boiling Point 78°C
Quantity 1 L
Explosion Limit Lower: 3.5 Vol %, Upper: 15 Vol %
Flash Point 12°C
Ignition Point 425°C
Odor Alcohol
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation Max. 0.001%
Titratable Acid Max. 0.0005 meq/g
Titratable Base Max. 0.0002 meq/g
Viscosity 1.2 mPas at 20°C
Water Max. 0.2%
Chemical Name or Material Ethanol, Absolute
Show More Show Less
compliance-icons
Product Identifier
  • Ethanol Absolute
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'