missing translation for 'onlineSavingsMsg'
Learn More

Ethanol absolute ≥99.5%, BAKER ANALYZED™, J.T. Baker™

Product Code. 15156996 Shop All J.T.Baker Products
Change view
Click to view available options
:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15156996 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 15156996 Supplier JT Baker Supplier No. 8029.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

For UV spectrophotometry

  • Ultraviolet Absorbance (1.00-cm path vs water; curve smooth throughout stated range with no extraneous impurity peaks):
    • at 210 nm max. 1.00
    • at 220 nm max. 0.50
    • at 230 nm max. 0.20
    • at 250 nm max. 0.05
    • at 270-400 nm max. 0.01

Specifications

CAS 64-17-5
Quantity 2.5 L
Assay Percent Range min. 99.5%
Residue after Evaporation max. 0.001%
Molecular Formula C2H6O
MDL Number MFCD00003568
Water max. 0.2%
Synonym Ethyl alcohol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.