missing translation for 'onlineSavingsMsg'
Learn More

Ethanol 99%+, Absolute, Extra Pure, SLR, Fisher Chemical™

Product Code. 10233962 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic Bottle
Plastic Container
Unit Size:
1 L
2.5 L
25 L
5 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10233962 25 L Metal Can 25 L
10680993 1 L Amber Glass Bottle 1 L
10048291 2.5 L Amber Glass Bottle 2.5 L
10610813 2.5 L HDPE Bottle 2.5 L
10375842 5 L Plastic Bottle 5 L
10385842 25 L Plastic Container 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 10233962 Supplier Fisher Chemical Supplier No. E/0600DF/25
  • ONLINE EXCLUSIVE PRICE
    £1510.11
    £1093.00 / 25 L
    Save £417.11
    27% Off
View all promotions
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Ethanol 99%+, Absolute, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
MDL Number 3568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Colorless
Quantity 25 L
Packaging Metal Can
Physical Form Liquid
Vapor Pressure 59 hPa at 20°C
Viscosity 1.2 mPaS at 20°C
Boiling Point 78°C
Molecular Weight (g/mol) 46.07
Formula Weight 46.07g/mol
compliance-icons compliance-icons
Product Identifier
  • Ethanol 99+%, absolute
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.