missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, 200 Proof (100%), USP/EP/ACS, Tax-Paid, Fisher Chemical™

Product Code. 15892534 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Glass Bottle
Steel Pail
Unit Size:
20 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15892534 20 L Steel Pail 20 L
15802544 4 L Glass Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15892534 Supplier Fisher Chemical Supplier No. A40920
  • ONLINE EXCLUSIVE PRICE
    £1290.00
    £1051.00 / 20 L
    Save £239.00
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 10-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

A tax-paid 200 proof ethanol that meets USP, EP, and ACS testing specifications to encourage cross-regional collaboration and potentially reduce laboratory inventory for ethanol applications.

  • Meets USP, EP, and ACS testing specifications
  • Consolidate your purchases into one product code and potentially reduce inventory
  • Suitable for use as a cleaner, solvent, or reagent in various applications
  • Packaged in laboratory-friendly sizes
  • Pricing includes tax
  • Combination of compendial test specifications enables use across regions
  • Intended for research use only; not for human or animal diagnostic purposes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Absorbance 0.40 max. at 240nm (USP, EP), 0.30 max at 250-260nm (USP, EP), 0.10 max at 270-340nm (USP, EP)
Color Colorless
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Residue after Evaporation 0.001% max. (ACS)
Specific Gravity null
Water 0.2% max. (ACS)
Boiling Point 78°C
Formula Weight 46.07
Grade ACS/EP/USP
Chemical Name or Material Ethanol, 200 Proof
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol 200 Proof USP EP ACS
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ethanol

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.