missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, 99.8%, anhyd.,denat. with 1%iPr-OH,1%MEK,0.001% Bitrex™

Product Code. 15835054 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2.5 L
5 L
Unit Size:
2.5 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15835054 5 L 5 L
15825054 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15835054 Supplier Thermo Scientific Supplier No. 463680050
  • ONLINE EXCLUSIVE PRICE
    £85.95
    £83.40 / 5 L
    Save £2.55
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64-17-5 | C2H6O | 46.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114.1°C
Density 0.7900 g/mL
Quantity 5 L
Flash Point 12°C
Assay Percent Range 99.8 to 100 % (GC)
Specific Gravity 0.79
Linear Formula C2H5OH
Boiling Point 78.0°C
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 46.06
Chemical Name or Material Ethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol, anhydrous, denaturated
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl alcohol, Methyl ethyl ketone, Isopropyl alcohol

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.