Learn More
Ethanethiol, 99+%, Thermo Scientific Chemicals
£14.90 - £140.00
Chemical Identifiers
CAS | 75-08-1 |
---|---|
Molecular Formula | C2H6S |
Molecular Weight (g/mol) | 62.13 |
MDL Number | MFCD00004887 |
InChI Key | DNJIEGIFACGWOD-UHFFFAOYSA-N |
Synonym | ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan |
PubChem CID | 6343 |
IUPAC Name | ethanethiol |
SMILES | CCS |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10306940
|
Thermo Scientific Acros
117860500 |
50 mL | Glass bottle |
£14.90
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10050800
|
Thermo Scientific Acros
117865000 |
500 mL | Glass bottle |
£40.90
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11340612
|
Thermo Scientific Acros
117860025 |
2.5 L | Glass bottle |
£140.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-08-1 | |
62.13 | |
DNJIEGIFACGWOD-UHFFFAOYSA-N | |
6343 | |
CCS |
C2H6S | |
MFCD00004887 | |
ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan | |
ethanethiol |
Specifications
75-08-1 | |
Colorless | |
35.0°C | |
Authentic | |
C2H6S | |
1.4296 to 1.4316 | |
01,34 | |
15,3781 | |
ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan | |
DNJIEGIFACGWOD-UHFFFAOYSA-N | |
ethanethiol | |
6343 | |
99+% | |
Ethanethiol, 99+% |
-148.0°C | |
0.8300g/mL | |
−45°C | |
99% min. (GC) | |
C2H5SH | |
MFCD00004887 | |
12,29 | |
0.83 | |
Solubility in water: 6.76g/L (20°C). Other solubilities: soluble in alcohol and ether | |
CCS | |
62.13 | |
62.13 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
Extremely flammable liquid and vapor.
GHS P Statement
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-837-3
missing translation for 'rtecsNumber' : KI9625000
missing translation for 'tsca' : TSCA
RUO – Research Use Only