missing translation for 'onlineSavingsMsg'
Learn More

Ethanesulfonic acid, 96%

Product Code. 11336489
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336489 250 g 250g
11327098 10 g 10g
11317098 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11336489 Supplier Thermo Scientific Alfa Aesar Supplier No. 014891.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 594-45-6 | C2H5O3S | 109.12 g/mol

Ethanesulfonic acid is employed in the electrolytic reduction of perrhenate solutions. It is also used as a catalyst for alkylation, polymerization and other reactions. Further, it reacts with triethoxymethane to prepare ethanesulfonic acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-45-6
Molecular Formula C2H5O3S
Molecular Weight (g/mol) 109.12
MDL Number MFCD00007529
InChI Key CCIVGXIOQKPBKL-UHFFFAOYSA-M
Synonym ethylsulfonic acid, ethanesulphonic acid, ethane sulfonate, ethane sulfonic acid, ethane sulphonic acid, ethansulfonic acid, ethylsulphonic acid, 1-ethanesulfonic acid, 2-ethanesulfonic acid, ethanesulfonicacid
PubChem CID 11668
ChEBI CHEBI:42465
IUPAC Name ethanesulfonic acid
SMILES CCS([O-])(=O)=O

Specifications

Melting Point -17°C
Color Colorless to Yellow
Density 1.343 g/mL
Boiling Point 122°C to 123°C (0.01 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.435
Quantity 250 g
UN Number UN2586
Beilstein 1743071
Solubility Information Miscible with water.
Formula Weight 110.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Ethanesulfonic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.