Learn More
Ethanesulfonic acid, 95%, ACROS Organics™
Brand: Acros Organics 366040050
Code : 29
Additional Details : Weight : 0.06000kg Transport : UN number : 2586 Chem class : 8 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
594-45-6 | |
110.13 | |
CCIVGXIOQKPBKL-UHFFFAOYSA-N | |
11668 | |
ethanesulfonic acid |
C2H6O3S | |
MFCD00007529 | |
ethylsulfonic acid, ethanesulphonic acid, ethane sulfonate, ethane sulfonic acid, ethane sulphonic acid, ethansulfonic acid, ethylsulphonic acid, 1-ethanesulfonic acid, 2-ethanesulfonic acid, ethanesulfonicacid | |
CHEBI:42465 | |
CCS(=O)(=O)O |
Specifications
Ethanesulfonic acid | |
>110°C | |
Glass bottle | |
1.343 | |
Yellow to Brown | |
5g | |
594-45-6 | |
C2H6O3S | |
MFCD00007529 | |
ethylsulfonic acid, ethanesulphonic acid, ethane sulfonate, ethane sulfonic acid, ethane sulphonic acid, ethansulfonic acid, ethylsulphonic acid, 1-ethanesulfonic acid, 2-ethanesulfonic acid, ethanesulfonicacid | |
CCIVGXIOQKPBKL-UHFFFAOYSA-N | |
ethanesulfonic acid | |
11668 | |
110.13 | |
95% |
1.3430g/mL | |
Authentic | |
1.4325 to 1.4355 | |
136°C to 140°C (3.0 hPa) | |
-17.0°C | |
95% | |
95% | |
C2H5SO3H | |
04, 5 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol | |
CCS(=O)(=O)O | |
110.13 | |
CHEBI:42465 | |
Liquid |
Safety and Handling
- Ethanesulfonic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.