missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethane-d5-thiol, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Ethane-d5-thiol |
| CAS | 61260-03-5 |
| Molecular Formula | C2 D5 H S |
| Quantity | 1 g |
| Synonym | Ethane-d5-thiol (9 CI), Ethanethiol-d5, Ethyl-d5 mercaptan, Thioethanol-d5, Ethanethiol-D5 |
| InChI Formula | InChI=1 S/C2H6S/c1-2-3/h3H,2H2,1H3/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])S |
| IUPAC Name | 1,1,2,2,2-pentadeuterioethanethiol |
| Molecular Weight (g/mol) | 67.1649 |
| Formula Weight | 67.0504 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction