missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethane-1,2-diol (Ethylene Glycol), Mikromol™
Description
Ethane-1,2-diol (Ethylene Glycol)
Specifications
Specifications
| Chemical Name or Material | Ethylene glycol |
| CAS | 107-21-1 |
| Analyte or Component Names | Ethylene glycol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C2 H6 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Hydroxyethyl Salicylate Imp. B (EP), Ethylene Glycol, Propan-1,2-diol, Glycerol Impurity B, Hydroxyethyl Salicylate Impurity B |
| InChI Formula | InChI=1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2 |
| SMILES | OCCO |
| IUPAC Name | ethane-1,2-diol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?