missing translation for 'onlineSavingsMsg'
Learn More

Ethane-1,2-diamine Mono(4-methylbenzenesulphonate), Mikromol™

Product Code. 17980906
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980906 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17980906 Supplier Mikromol Supplier No. MM0089.080025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethane-1,2-diamine Mono(4-methylbenzenesulphonate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethane-1,2-diamine Mono(4-methylbenzenesulphonate)
CAS 14034-59-4
Analyte or Component Names Ethane-1,2-diamine Mono(4-methylbenzenesulphonate)
Shipping Condition Room Temperature
Molecular Formula C7 H8 O3 S . C2 H8 N2
Certifications/Compliance ISO 9001
Synonym 1,2-Ethanediamine, 4-methylbenzenesulfonate (1:1), 1,2-Ethanediamine, mono(4-methylbenzenesulfonate) (9CI), Ethylenediamine, mono-p-toluenesulfonate (8CI), 2-Aminoethylammonium p-toluenesulfonate, Ethylenediamine monotosylate, Ethylenediamine p-toluenesulfonate, NSC 14086, Xylometazoline Hydrochloride Impurity E, Xylometazoline Impurity E
InChI Formula InChI=1S/C7H8O3S.C2H8N2/c1-6-2-4-7(5-3-6)11(8,9)10;3-1-2-4/h2-5H,1H3,(H,8,9,10);1-4H2
SMILES Cc1ccc(cc1)S(=O)(=O)O.NCCN
IUPAC Name ethane-1,2-diamine;4-methylbenzenesulfonic acid
Molecular Weight (g/mol) 232.2999
Quantity 25 mg
Formula Weight 232.0882 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.