missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethametsulfuron-methyl Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Ethametsulfuron-methyl Sodium Salt |
| Molecular Formula | C15 H17 N6 Na O6 S |
| Quantity | 50 mg |
| Synonym | DPX-A 7881 Sodium Salt, Ethametsulfuron Methyl Ester Sodium Salt, Ethametsulfuron-methyl Sodium Salt, Muster Sodium Salt, Salsa Sodium Salt, Salsa 75WG Sodium Salt, 2-[[[[[4-Ethoxy-6-(methylamino)-1,3,5-triazin-2-yl]amino]carbonyl]amino]sulfonyl]-benzoic Acid Methyl Ester Sodium Salt |
| InChI Formula | InChI=1S/C15H18N6O6S.Na/c1-4-27-15-19-12(16-2)17-13(20-15)18-14(23)21-28(24,25)10-8-6-5-7-9(10)11(22)26-3;/h5-8H,4H2,1-3H3,(H3,16,17,18,19,20,21,23);/q;+1/p-1 |
| SMILES | CCOc1nc(NC)nc(NC(=O)N([Na])S(=O)(=O)c2ccccc2C(=O)OC)n1 |
| IUPAC Name | [[4-ethoxy-6-(methylamino)-1,3,5-triazin-2-yl]carbamoyl-(2-methoxycarbonylphenyl)sulfonylamino]sodium |
| Molecular Weight (g/mol) | 432.387 |
| Formula Weight | 432.083 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?