missing translation for 'onlineSavingsMsg'
Learn More

Ethambutol Dihydrochloride, TRC

Product Code. 30007551
Change view
Click to view available options
Quantity:
50 mg
5 g
25 g
Unit Size:
25g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007551 25 g 25g
18091540 5 g 5g
18092130 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007551 Supplier Toronto Research Chemicals Supplier No. TRCE88980025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-11-7
Molecular Formula C10 H24 N2 O2 . 2 Cl H
Molecular Weight (g/mol) 277.23
Synonym Ethambutol hydrochloride, (2S,2'S)-2,2'-(Ethylenediimino)dibutan-1-ol dihydrochloride, 1-Butanol, 2,2'-(1,2-ethanediyldiimino)bis-, hydrochloride (1:2), (2S,2'S)-, 1-Butanol, 2,2'-(1,2-ethanediyldiimino)bis-, dihydrochloride, (2S,2'S)- (9CI), 1-Butanol, 2,2'-(1,2-ethanediyldiimino)bis-, dihydrochloride, [S-(R*,R*)]-, 1-Butanol, 2,2'-(ethylenediimino)di-, dihydrochloride, (+)- (8CI), (+)-2,2'-(Ethylenediimino)di-1-butanol dihydrochloride, CL 40881, Dadibutol, Dexambutol, Ebutol, Etapiam, Ethambutol dihydrochloride, Ethambutol hydrochloride, Ethambutold dihydrochloride, Etibi, Myambutol, Mycobutol, Sural, Tambutol, Tibutol, d-2,2'-(Ethylenediimino)-di-1-butanol dihydrochloride
IUPAC Name (2S)-2-[2-[[(2S)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol;dihydrochloride
SMILES Cl.Cl.CC[C@@H](CO)NCCN[C@@H](CC)CO

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H24N2O2.2ClH/c1-3-9(7-13)11-5-6-12-10(4-2)8-14;;/h9-14H,3-8H2,1-2H3;2*1H/t9-,10-;;/m0../s1
Formula Weight 276.1371
Percent Purity >95%
Chemical Name or Material Ethambutol hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.