Learn More
Ethambutol dihydrochloride, Thermo Scientific™
An antibacterial agent
£142.00
Chemical Identifiers
| CAS | 1070-11-7 |
|---|---|
| Molecular Formula | C10H26Cl2N2O2 |
| Molecular Weight (g/mol) | 277.23 |
| MDL Number | MFCD00216025 |
| InChI Key | AUAHHJJRFHRVPV-HSTMFJOWSA-N |
| Synonym | ethambutol 2hcl, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethyl amino butan-1-ol dihydrochloride, ethambutol related compound b usp, prestwick_208, aronis24117, 2,2'-1,2-ethanediyldiimino bis-1-butanol dihydrochloride, unii-jng307dj5x component, 2,2'-1,2-ethanediyldi imino di 1-butanol dihydrochloride, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethylamino butan-1-ol dihydrochloride, 2r,2'r-2,2'-ethane-1,2-diylbis azanediyl bis butan-1-ol dihydrochloride |
| PubChem CID | 6419929 |
| IUPAC Name | (2R)-2-[2-[[(2R)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol;dihydrochloride |
| SMILES | CCC(CO)NCCNC(CC)CO.Cl.Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySlightly soluble in water
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| C10H26Cl2N2O2 | |
| MFCD00216025 | |
| ethambutol 2hcl, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethyl amino butan-1-ol dihydrochloride, ethambutol related compound b usp, prestwick_208, aronis24117, 2,2'-1,2-ethanediyldiimino bis-1-butanol dihydrochloride, unii-jng307dj5x component, 2,2'-1,2-ethanediyldi imino di 1-butanol dihydrochloride, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethylamino butan-1-ol dihydrochloride, 2r,2'r-2,2'-ethane-1,2-diylbis azanediyl bis butan-1-ol dihydrochloride | |
| (2R)-2-[2-[[(2R)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol;dihydrochloride |
Specifications
| 1070-11-7 | |
| MFCD00216025 | |
| ethambutol 2hcl, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethyl amino butan-1-ol dihydrochloride, ethambutol related compound b usp, prestwick_208, aronis24117, 2,2'-1,2-ethanediyldiimino bis-1-butanol dihydrochloride, unii-jng307dj5x component, 2,2'-1,2-ethanediyldi imino di 1-butanol dihydrochloride, 2r-2-2-2r-1-hydroxybutan-2-yl amino ethylamino butan-1-ol dihydrochloride, 2r,2'r-2,2'-ethane-1,2-diylbis azanediyl bis butan-1-ol dihydrochloride | |
| AUAHHJJRFHRVPV-HSTMFJOWSA-N | |
| (2R)-2-[2-[[(2R)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol;dihydrochloride | |
| 6419929 | |
| Ethambutol dihydrochloride |
| C10H26Cl2N2O2 | |
| 14,3720 | |
| Slightly soluble in water | |
| CCC(CO)NCCNC(CC)CO.Cl.Cl | |
| 277.23 | |
| 277.23 |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H360
missing translation for 'einecsNumber' : 213-970-7
missing translation for 'rtecsNumber' : EL3854000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.