missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethambutol dihydrochloride, MedChemExpress
Description
Ethambutol dihydrochloride (Emb dihydrochloride) is a bacteriostatic antimycobacterial agent, which obstructs the formation of cell wall by inhibiting arabinosyl transferases.
Specifications
Specifications
| Chemical Name or Material | Ethambutol dihydrochloride |
| Physical Form | Solid |
| CAS | 1070-11-7 |
| Color | White |
| Quantity | 1 g |
| Purity Grade Notes | Research |
| Molecular Formula | C10H26Cl2N2O2 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | Emb dihydrochloride |
| Solubility Information | DMSO : 100 mg/mL (360.71 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (180.36 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?