missing translation for 'onlineSavingsMsg'
Learn More

Ethambutol dihydrochloride, MedChemExpress

Product Code. 16410988
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16410988

Brand: Medchem Express HYB0535A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ethambutol dihydrochloride (Emb dihydrochloride) is a bacteriostatic antimycobacterial agent, which obstructs the formation of cell wall by inhibiting arabinosyl transferases.

Ethambutol dihydrochloride (Emb dihydrochloride) is a bacteriostatic antimycobacterial agent, which obstructs the formation of cell wall by inhibiting arabinosyl transferases.

Specifications

Chemical Name or Material Ethambutol dihydrochloride
Physical Form Solid
CAS 1070-11-7
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C10H26Cl2N2O2
For Use With (Application) COVID-19-anti-virus
Synonym Emb dihydrochloride
Solubility Information DMSO : 100 mg/mL (360.71 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (180.36 mM)
SMILES CC[C@H](NCCN[C@@H](CC)CO)CO.Cl.Cl
Molecular Weight (g/mol) 277.23
Formula Weight 277.23
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.