missing translation for 'onlineSavingsMsg'
Learn More

Ethacrynic acid

Product Code. 15405869
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15405869 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15405869 Supplier Thermo Scientific Alfa Aesar Supplier No. J63684.03
  • ONLINE EXCLUSIVE PRICE
    £67.30
    £60.65 / 1g
    Save £6.65
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58-54-8 | C13H12Cl2O4 | 303.135 g/mol

Ethacrynic acid is used to inhibits symport of sodium, potassium, and chloride primarily in the ascending limb of Henle, but also in the proximal and distal tubules. This pharmacological action results in excretion of these ions, increased urinary output, and reduction in extracellular fluid. This compound has been classified as a loop or high ceiling diuretic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethacrynic acid is used to inhibits symport of sodium, potassium, and chloride primarily in the ascending limb of Henle, but also in the proximal and distal tubules. This pharmacological action results in excretion of these ions, in
TRUSTED_SUSTAINABILITY

Specifications

CAS 58-54-8
Color White
Molecular Formula C13H12Cl2O4
MDL Number MFCD00056693
Quantity 1 g
Merck Index 14,3717
Synonym ethacrynic acid, etacrynic acid, ethacrynate, etacrinic acid, hydromedin, edecrin, crinuryl, edecril, edecrina, endecril
Solubility Information Soluble in ethanol,chloroform,ether,ammonia,carbonates,and methanol. Insoluble in water.
InChI Key AVOLMBLBETYQHX-UHFFFAOYSA-N
SMILES CCC(=C)C(=O)C1=C(C(=C(C=C1)OCC(=O)O)Cl)Cl
IUPAC Name 2-[2,3-dichloro-4-(2-methylidenebutanoyl)phenoxy]acetic acid
Molecular Weight (g/mol) 303.135
PubChem CID 3278
ChEBI CHEBI:4876
Formula Weight 303.14
Physical Form Crystalline Powder
Chemical Name or Material Ethacrynic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.