missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ ETFE Thread Adapter

Product Code. 10111291
Click to view available options
Size:
28 mm
32 mm
38 mm
38/430 mm
40 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10111291

Brand: Hirschmann™ 9313621

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Threaded adapter ETFE for use with ceramus™, solarus™ and opus™.

  • For the adaption on the most common bottles

Specifications

Diameter (Metric) Inner 45 mm
Type Thread Adapter
Material ETFE
Size 32 mm
For Use With (Equipment) ceramus™ and opus™
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.