Learn More
BRAND™ ETFE Bottle Adapter
For use with Dispensette™ seripettor™, Titrette™ and QuikSip™ bottle-top dispensers.
£17.90 - £24.55
Specifications
Material | ETFE |
---|---|
For Use With (Equipment) | Dispensette™, seripettor™, Titrett™, QuikSip™ |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
10787021
|
BRAND™
704375 |
GL 24 - 25 |
£17.90
Each |
In Stock
Log in to see stock. |
|||||
10612641
|
BRAND™
704378 |
GL 28 |
£17.90
Each |
In Stock
Log in to see stock. |
|||||
10576522
|
BRAND™
704398 |
GL 32 - 33 |
£24.55
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Description
- ETFE Construction
- For use with Dispensette™ seripettor™ Titrette™ and QuikSip™ Bottle-top dispensers
Specifications
ETFE | |
Dispensette™, seripettor™, Titrett™, QuikSip™ |