Learn More
DWK Life Sciences ETFE Sealing Plug 1/4 in. - 28 Thread
ETFE sealing plug for 3-port HPLC GL 45 delivery cap
Brand:  DWK Life Sciences 292310003
6.07 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 15859007
  • £6.65 / Each
Estimated Shipment: 15-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
ETFE Sealing Plug 1/4 in. - 28 thread for 3-port HPLC GL 45 delivery cap
  • chemically resistant
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Supply of solvents to HPLC equipment; Safe transfer of liquid media within a closed and sterile system; Chemical and thermal reactions; Storage and transport of media, APIs and reagents; Transport and shipping
ETFE
1/4 in. - 28