missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETFDH Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559808
This item is not returnable.
View return policy
Description
Immunogen sequence: LAVAHEKDIR VCLVEKAAQI GAHTLSGACL DPGAFKELFP DWKEKGAPLN TPVTEDRFGI LTEKYRIPVP ILPGLPMN Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
Electron-transferring-flavoprotein dehydrogenase in the inner mitochondrial membrane accepts electrons from electron-transfer flavoprotein which is located in the mitochondrial matrix and reduces ubiquinone in the mitochondrial membrane. The protein is synthesized as a 67-kDa precursor which is targeted to mitochondria and processed in a single step to a 64-kDa mature form located in the mitochondrial membrane. Deficiency in electron-transferring-flavoprotein dehydrogenase have been demonstrated in some patients with type II glutaricacidemia.
Specifications
| ETFDH | |
| Polyclonal | |
| Unconjugated | |
| Etfdh | |
| 0610010I20Rik; AV001013; electron transfer flavoprotein dehydrogenase; electron transfer flavoprotein-ubiquinone oxidoreductase, mitochondrial; electron transferring flavoprotein dehydrogenase; electron transferring flavoprotein, dehydrogenase; electron-transferring-flavoprotein dehydrogenase; ETF dehydrogenase; Etfdh; ETFQO; ETF-QO; ETF-ubiquinone oxidoreductase; MADD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2110 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16134 | |
| Etfdh | |
| Recombinant protein corresponding to Human ETFDH. Recombinant protein control fragment (Product #RP-96739). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction