missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETFB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106327
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ETFB.
This gene encodes electron-transfer-flavoprotein, beta polypeptide, which shuttles electrons between primary flavoprotein dehydrogenases involved in mitochondrial fatty acid and amino acid catabolism and the membrane-bound electron transfer flavoprotein ubiquinone oxidoreductase. The gene deficiencies have been implicated in type II glutaricaciduria. Alternatively spliced transcript variants have been found for this gene.
Specifications
| ETFB | |
| Polyclonal | |
| Unconjugated | |
| ETFB | |
| 0610009I16Rik; 2810441H06Rik; beta-ETF; electron transfer flavoprotein beta subunit; electron transfer flavoprotein beta-subunit; electron transfer flavoprotein subunit beta; electron transfer flavoprotein, beta polypeptide; electron transferring flavoprotein, beta polypeptide; electron-transfer-flavoprotein, beta polypeptide; electron-transferring-flavoprotein, beta polypeptide; Etfb; FP585; MADD | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 110826, 2109, 292845 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P38117, Q68FU3, Q9DCW4 | |
| ETFB | |
| A synthesized peptide derived from human ETFB(Accession P38117), corresponding to amino acid residues A105-L155. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction