missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETFB Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-49843
This item is not returnable.
View return policy
Description
ETFB Polyclonal specifically detects ETFB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Peptide ELISA.
Specifications
| ETFB | |
| Polyclonal | |
| Western Blot 1 - 3 ug/ml, Immunohistochemistry 3 - 5 ug/ml, Peptide ELISA Detection limit 1:8000 | |
| NP_001976.1 | |
| ETFB | |
| Peptide with sequence C-QVTLEGDKLKVERE corresponding to internal region according to NP_001014763.1. | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 2109 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| beta-ETF, electron transfer flavoprotein beta subunit, electron transfer flavoprotein beta-subunit, electron transfer flavoprotein subunit beta, electron transfer flavoprotein, beta polypeptide, electron-transfer-flavoprotein, beta polypeptide, electron-transferring-flavoprotein, beta polypeptide, MADD | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_001976.1; NP_001014763.1). Amino acid numbering in the name above refers to NP_001976.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction