Learn More
Invitrogen™ ETFA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530021
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, C8D30, PC-12. Predicted reactivity: Mouse (96%), Rat (95%), Zebrafish (84%), Xenopus laevis (85%), Cat (96%), Pig (96%), Chicken (87%), Rhesus Monkey (98%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ETFA participates in catalyzing the initial step of the mitochondrial fatty acid beta-oxidation. It shuttles electrons between primary flavoprotein dehydrogenases and the membrane-bound electron transfer flavoprotein ubiquinone oxidoreductase. Defects in electron-transfer-flavoprotein have been implicated in type II glutaricaciduria in which multiple acyl-CoA dehydrogenase deficiencies result in large excretion of glutaric, lactic, ethylmalonic, butyric, isobutyric, 2-methyl-butyric, and isovaleric acids. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| ETFA | |
| Polyclonal | |
| Unconjugated | |
| ETFA | |
| 2010200I21Rik; Alpha-ETF; D9Ertd394e; electron transfer flavoprotein alpha subunit; electron transfer flavoprotein alpha-subunit; electron transfer flavoprotein subunit alpha, mitochondrial; electron transfer flavoprotein, alpha polypeptide; electron transferring flavoprotein, alpha polypeptide; electron-transfer-flavoprotein alpha polypeptide; electron-transfer-flavoprotein, alpha polypeptide; EMA; ETF; ETFA; GA2; glutaric aciduria II; MADD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 110842, 2108, 300726 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.91 mg/mL | |
| PBS with 20% glycerol; pH 7 | |
| P13803, P13804, Q99LC5 | |
| ETFA | |
| Recombinant protein encompassing a sequence within the center region of human ETFA. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.