missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10884-1-AP-150UL
This item is not returnable.
View return policy
Description
Termination of protein biosynthesis and release of the nascent polypeptide chain are signaled by the presence of an in-frame stop codon at the aminoacyl site of the ribosome. The process of translation termination is universal and is mediated by protein release factors.Specifications
| ETF1 | |
| Polyclonal | |
| Unconjugated | |
| ETF1 | |
| D5S1995, ERF, ERF1, ETF1, Eukaryotic release factor 1, Protein Cl1, RF1, SUP45L1, TB3 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2107, 225363 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P62495, Q8BWY3 | |
| ETF1 | |
| ETF1 Fusion Protein Ag1330 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction